Starch, soluble, powder ACS suitable for use as excipient
Supplier: SPECTRUM CHEMICAL MFG CORP CA
Synonyms:
Starch, Vitex® iodine indicator, Amylodextrin
Starch, Soluble, Powder, Reagent, ACS is indicator for redox titrations where triiodide is present. Starch forms a very dark blue-black complex with triiodide which can be made by mixing iodine with iodide (often from potassium iodide). The color of the starch complex is so deep, that it can be detected visually when the concentration of the iodine is as low as 0.00002 M at 20 °C.
As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.
Formula:
(C₆H₁₀O₅)n MW: 162.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00082026 CAS Number: 9005-84-9 EINECS: 232-686-4 Merck Index: 13,08878 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...