Sodium sulfite 98 ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP CA

S1475-12KG S1475-2.5KG S1475-500GM
CA700001-458EA 2429 CAD
CA700001-458 CA700001-460 CA700001-464
Sodium sulfite 98 ACS for synthesis
Sodium sulphite

Sodium Sulfite, Reagent, ACS is a used for preserving meat and prevent discoloration of dried fruit. It is a soluble sodium salt of sulfuric acid. It is also used in the paper processing industry and to purify TNT. As an ACS grade Reagent, Spectrum Chemical manufactured compound is used as the quality standard against which other substances are grade and has met the toughest regulatory standards for quality and pureness

Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: > 500 °C
Density: 2.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR