Sodium gluconate ≥98% USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: D-Gluconic acid sodium salt, Sodium (2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoate, Gluconic acid, sodium salt, Sodium D-gluconate

S1286-12KG S1286-500GM
CA700011-916EA 3722.01 CAD
CA700011-916 CA95035-822
Sodium gluconate ≥98% USP suitable for use as excipient
Sodium gluconate

Sodium Gluconate, USP is used as surface cleaning agent for metals and glass bottles. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HOH₂C(CH(OH))₄COONa
MW: 218.14 g/mol
Melting Pt: 170…175 °C
Storage Temperature: Ambient
MDL Number: MFCD00064210
CAS Number: 527-07-1
EINECS: 208-407-7
Merck Index: 12,08766
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eafd5b786cb23fb5c20dd64ad056ed2d80dc0186&vid=b1ffe48dafca4b8bd9b0d709ea3e986099051767&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR