Polysorbate 80 NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate

PO138-20LT PO138-4LTGL PO138-7GLBL
CA700011-064EA 4095 CAD
CA700011-064 CA71002-434 75812-046 75812-048
Polysorbate 80 NF suitable for use as excipient
Polysorbate 80

Polysorbate 80, NF is used to solubilize essential oils.


  • BSE/TSE statement (available upon request)
  • Allergen statement (available upon request)
  • Kosher certified


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


Caution: Storage temperature - Keep tightly closed, protect from light.

Melting Pt: 100 °C
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR