Polyvinylpyrrolidone, M.W. ∼ 40,000 K-30 USP

Supplier: Spectrum Chemical Mfg. Corp.

P1454-12KGBL P1454-50KGBL
CA89078-978EA 14346.01 CAD
CA89078-978 CA89078-980
Polyvinylpyrrolidone, M.W. ∼ 40,000 K-30 USP
Polyvinylpyrrolidone

Melamine statement (available upon request) BSE/TSE statement (available upon request)


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: (C₆H₉NO)n
Storage Temperature: Ambient
MDL Number: MFCD01076626
CAS Number: 9003-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR