Potassium thiocyanate ≥98.5%, crystals ACS
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Potassium rhodanide
Potassium Thiocyanate, Crystal, Reagent, ACS, one of the pseudohalides, is an important salt of the thiocyanate anion. Often used in chemical synthesis, it can also be used in theatrical special effects to create a relatively realistic blood effect. As an ACS grade Reagent, Spectrum Chemical manufactured potassium thiocyanate is used as the quality standard against which other substances are graded and has met the toughest regulatory standards for quality and pureness
Formula:
KSCN MW: 97.18 g/mol Boiling Pt: 500 °C (1013 hPa) Melting Pt: 175 °C Density: 1.61 g/cm³ (20 °C) |
MDL Number:
MFCD00011413 CAS Number: 333-20-0 Merck Index: 12,07866 |
Specification Test Results
Assay | 98.5% Min. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...