Polysorbate 20 FCC
Supplier: Spectrum Chemical Mfg. Corp.
Polysorbate 20, FCC is used as a wetting agent. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Allergen statement (available upon request)
Formula:
C₅₈H₁₁₄O₂₆ Density: 1.095 Storage Temperature: Ambient |
MDL Number:
MFCD00165986 CAS Number: 9005-64-5 EINECS: 500-018-3 |
Specification Test Results
Assay | 70% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...