Mineral oil heavy USP suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP CA
Warning

MI500-20LTPL
CA89078-896EA 2000 CAD
CA89078-896
Mineral oil heavy USP suitable for use as excipient
Mineral oil

Mineral Oil, Heavy, USP is a lubricant excipient material. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Boiling Pt: 300 °C (1013 hPa)
Density: 0.872 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8012-95-1
EINECS: 232-455-8

Order Now

Specification Test Results

SPECIFIC GRAVITY @ 25oC 0.845-0.905
VISCOSITY @40.0 C 34.5-150.0 mm²


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR