Mineral oil, white (petroleum) light NF
Supplier: SPECTRUM CHEMICAL MFG CORP CA
|
Warning
|
M1293-20LT
CA89078-900EA
3818.01
CAD
CA89078-900
Mineral oil, white (petroleum) light NF
White mineral oil (petroleum)
Light Mineral Oil, NF is used as a vehicle and humectant.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Boiling Pt:
300 °C (1013 hPa) Density: 0.872 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00131611 CAS Number: 8042-47-5 EINECS: 232-455-8 |
Specification Test Results
SPECIFIC GRAVITY @ 25°C | 0.818-0.880 |
VISCOSITY @40.0 °C | 3.0-34.4 mm²/s |
ACIDITY | TO PASS TEST |
READILY CARBONIZABLE SUBSTANCES | TO PASS TEST |
POLYCYCLIC AROMATIC HYDROCARBONS | TO PASS TEST |
SOLID PARAFFIN | TO PASS TEST |
SULFUR COMPOUNDS | TO PASS TEST |
ELEMENTAL IMPURITIES | AS REPORTED |
IDENTIFICATION | TO PASS TEST |
RESIDUAL SOLVENTS | TO PASS TEST |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...