Magnesium sulfate, anhydrous ≥99%, powder, Reagent Grade
Supplier: Spectrum Chemical Mfg. Corp.
M1063-125GM
M1063-12KG
M1063-2.5KG
M1063-500GM
M1063-50KGBL
CA700001-502EA
298
CAD
CA700001-502
CA700001-504
CA700001-506
CA700001-508
CA700001-510
Magnesium sulfate, anhydrous ≥99%, powder, Reagent Grade
Magnesium sulphate
Magnesium Sulfate, Anhydrous, Powder, Reagent is the raw material for a common magnesium-based, called Epsom salts. The Epsom salt formulations are for both topical and oral indications. The topical preparation is a gel for soothing aches and pains while the internal formulation is commonly used as saline laxative. In it's anhydrous form, magnesium sulfate is used as a drying agent.
Formula:
MgSO₄ Melting Pt: 1127 °C Density: 2.65 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011110 CAS Number: 7487-88-9 EINECS: 231-298-2 Merck Index: 12,05731 |
|
Specification Test Results
Assay | 99% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...