Lanthanum(III) chloride, hydrated ACS for synthesis

Supplier: SPECTRUM CHEMICAL MFG CORP
Warning

Synonyms: Lanthanum chloride, Lanthanum trichloride

L1021-25GM L1021-125GM
700002-102EA 209.18 CAD
700002-102 700002-100
Lanthanum(III) chloride, hydrated ACS for synthesis
Lanthanum (III) chloride

Lanthanum Chloride, Hydrated, Reagent, ACS, is used in biochemical research to block the activity of divalent cation channels, mainly calcium channels. Doped with cerium, it is used as a scintillator material and in organic synthesis, lanthanum trichloride functions as a mild Lewis acid for converting aldehydes to acetals. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: Cl₃La
MW: 245.26 g/mol
Melting Pt: 857
Density: 3.8773 g/cm³ (–0.15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011068
CAS Number: 10099-58-8
EINECS: 233-237-5
Merck Index: 13,05379

Order Now

Specification Test Results

Assay 64.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR