L(+)-Glutamine ≥98.5% FCC suitable for use as excipient, for biochemistry

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Gln, L-Glutamine, H-Gln-OH, (S)-(+)-Glutamine, (S)-2,5-Diamino-5-oxopentanoic acid, Levoglutamide, Q

G1039-1KG G1039-25GM G1039-12KGBL G1039-100GM G1039-12KG G1039-50KGBL
95033-950EA 1678.21 CAD
95033-950 95033-952 700007-758 95033-948 89050-434 89050-436
L(+)-Glutamine ≥98.5% FCC suitable for use as excipient, for biochemistry
L(+)-Glutamine

L-Glutamine, FCC - The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₅H₁₀N₂O₃
Boiling Pt: 355 °C (1013 hPa)
Melting Pt: 185…186 °C
Density: 1.5326 g/cm³ (130 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008044
CAS Number: 56-85-9
EINECS: 200-292-1
Merck Index: 13,04484
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cb7a812ec5d2c73a797fc464926df9eef12c5c9a&vid=15d7d87daff0fec33a974a578415e157a3278049&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR