D-(-)-Fructose ≥98%, granular USP

Supplier: Spectrum Chemical Mfg. Corp.

FR106-45KGBL FR106-12KG FR106-2.5CS4 FR106-2.5KG FR106-500CS6 FR106-500GM
CA700009-646EA 5867 CAD
CA700009-646 CA700009-644 CA95033-332 CA95033-334 CA95033-336 CA95033-338
D-(-)-Fructose ≥98%, granular USP
D(-)-Fructose

Fructose, Granular, USP is a white, odorless crystalline solid that is extremely sweet and is the most water soluble of all sugars.


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: C₆H₁₂O₆
Boiling Pt: 552 °C (1013 hPa)
Melting Pt: 100…110 °C
Density: 1.59 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00148910
CAS Number: 57-48-7
EINECS: 200-333-3
Merck Index: 13,04295
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8c96c34b23753d10b06108938876644c747995c&vid=6d952afc3154d3befcf243a015f513a9130c0835&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR