EDTA (ethylenediamine tetraacetic acid) ≥98% NF

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Ethylenediaminetetraacetic acid2,2',2'',2'''-(Ethane-1,2-diylbis(azanetriyl))tetraacetic acidOmniPur® EDTA(Ethylenedinitrilo)tetraacetic acidIdranal® IIKestranal® AEdetic acidEthylenedinitrilotetra-acetic acid

E1208-100GM E1208-10 E1208-25KGBL E1208-500GM
CA11024-328EA 1274.62 CAD
CA11024-328 700007-312 95033-074 95033-076
EDTA (ethylenediamine tetraacetic acid) ≥98% NF
EDTA (Ethylenediamine tetraacetic acid)

Edetic Acid, NF is used as a chelating agent in therapy and as preservative adjuncts. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: C₁₀H₁₆N₂O₈
MW: 292.25 g/mol
Boiling Pt: 614 °C (1013 hPa)
Melting Pt: 220 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: >100 °C
MDL Number: MFCD00003541
CAS Number: 60-00-4
Merck Index: 13,03546
https://ca.vwr.com/stibo/web/std.lang.all/72/41/2297241.jpg

Order Now

Specification Test Results

Assay 98% Min.

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR