Dextran, M.W. ~ 250,000, powder

Supplier: SPECTRUM CHEMICAL MFG CORP

DE130-25GM DE130-1KG DE130-100GM
700013-134EA 551.12 CAD
700013-134 700013-136 700013-138
Dextran, M.W. ~ 250,000, powder
Dextran

Dextran, 250,000, Powder is a complex branched glucan that is synthesized by certain lactic acid bacteria from sucrose. Commonly used in microsurgery to decrease the risk of vascular thrombosis, these agents are also used as effective osmotic agents and can treat hypovolemia. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules.

Formula: (C₆H₁₀O₅)n
Storage Temperature: Ambient
MDL Number: MFCD00130935
CAS Number: 9004-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR