Sodium dodecylbenzenesulfonate ≥90%, powder suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP
Danger

Synonyms: Laurylbenzenesulphonic acid Sodium Salt, Alkylbenzenesulphonic acid Sodium Salt, Sodium Laurylbenzenesulfonate, Sodium Alkylbenzenesulfonate, Laurylbenzenesulfonic acid Sodium Salt, Sodium Alkylbenzenesulphonate, Dodecylbenzenesulphonic acid sodium salt, Dodecylbenzenesulfonic acid sodium salt, Alkylbenzenesulfonic acid Sodium Salt, Sodium Laurylbenzenesulphonate

D1064-12KG D1064-500GM
700002-476EA 869.81 CAD
700002-476 CA11024-316
Sodium dodecylbenzenesulfonate ≥90%, powder suitable for use as excipient
Sodium dodecylbenzenesulphonate

Dodecylbenzene Sodium Sulfonate, Powder is a surfactant used in laboratory research.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₈H₂₉NaO₃S
Melting Pt: > 300 °C
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011508
CAS Number: 25155-30-0
EINECS: 246-680-4
Merck Index: 13,08686

Order Now

Specification Test Results

Assay 90% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR