Dextran, M.W. ~ 500,000, powder
Supplier: Spectrum Chemical Mfg. Corp.
D1004-100GM
D1004-1KG
D1004-25GM
CA700013-094EA
859
CAD
CA700013-094
CA700013-096
CA700013-098
Dextran, M.W. ~ 500,000, powder
Dextran
Dextran, 500,000, Powder is a complex branched glucan that is synthethized by certain lactic acid bacteria from sucrose. In the laboratory, Dextran is used to apply osmotic pressure to biological molecules and can, when applied with a fluorescent molecule, be used to create concentration gradients of diffusible molecules
Formula:
(C₆H₁₀O₅)n Storage Temperature: Ambient |
MDL Number:
MFCD00130935 CAS Number: 9004-54-0 EINECS: 232-677-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...