Choline chloride ≥98% FCC suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: 2-hydroxy-N,N,N-trimethylethanaminium chloride

CH147-50KGBL CH147-12KG CH147-500GMP
700007-522EA 3709.05 CAD
700007-522 700007-518 700007-520
Choline chloride ≥98% FCC suitable for use as excipient
Choline chloride

Choline Chloride, FCC is an essential nutrient and typically grouped with B-complex vitamins. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: HOH₂CCH₂N(CH₃)₃Cl
Melting Pt: 244…247 °C
Density: ∼1.205 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011721
CAS Number: 67-48-1
EINECS: 200-655-4
Merck Index: 13,02226
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=77b015e76f45084bc46da3249578a85652cd6802&vid=85c8a0a1b152b861b3bc3ec8fe25a70509221d85&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR