Cholesterol NF

Supplier: Spectrum Chemical Mfg. Corp.

CH135-125GM CH135-12KG CH135-2.5KG CH135-25GM CH135-500GM
CA700003-080EA 1062 CAD
CA700003-080 CA700003-082 CA700003-084 CA700003-086 CA700003-088
Cholesterol NF
Cholesterol
Formula: C₂₇H₄₆O
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 147…150 °C
Density: 1.07 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003646
CAS Number: 57-88-5
EINECS: 200-353-2
Merck Index: 13,02221

Order Now

Specification Test Results

ASSAY (DRIED BASIS) 95.0 - 102.0 %
MELTING RANGE 147° - 150°C
SPECIFIC ROTATION -34° to -38°
SOLUBILITY IN ALCOHOL No deposit or turbidity forms
ACIDITY max 0.3 ml
LOSS ON DRYING max 0.3%
RESIDUE ON IGNITION max 0.1%
RELATED STEROLS AND OTHER ORGANIC IMPURITIES:
β-CHOLESTANOL (5a-CHOLESTAN-3β-ol,DIHYDROCHOLESTEROL) max 0.6 %
DESMOSTEROL max 4%
LATHOSTEROL max 2%
24-DEHYDROLATHOSTEROL (5a-CHOLESTA-7,24 DIEN--3β-ol ) max 0.2 %
4-METHYLCHOLEST-5-EN-3β-OL max 0.5 %
TOTAL IMPURITIES INCLUDING RELATED STEROLS max 5%
ELEMENTAL IMPURITIES AS REPORTED
IDENTIFICATION (A) SPECTRUM MATCHES REFERENCE
IDENTIFICATION (B) -34° to -38°
IDENTIFICATION (C) 147 - 150 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR