Carboxymethyl cellulose sodium salt, 400-800 cP (2% H₂O - 25°C) USP, medium viscosity

Supplier: SPECTRUM CHEMICAL MFG CORP CA

Synonyms: Carboxymethyl cellulose, sodium saltCellulose, carboxymethyl ether, sodium saltCMCCellulose gumSodium carboxymethyl celluloseSodium carboxymethyl cellulose (Viscosity 800mpa.s)

CA192-2.5KG CA192-125GM CA192-500GM CA192-45KGBL CA192-45KGBL CA192-12KG
CA89078-698EA 33733.46 CAD
CA89078-698 89078-698 95032-434 95032-438 89078-696 95032-436
Carboxymethyl cellulose sodium salt, 400-800 cP (2% H₂O - 25°C) USP, medium viscosity
Carboxymethyl cellulose sodium salt

Carboxymethylcellulose Sodium, Medium Viscosity, Viscosity of 2 Percent Aqueous Solution @ 25 DEG C: 400-800 cP, USP is often used as a thickener and stabilizer. 


All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formula: [C₆H₇O₂(OH)x(OCH₂COONa)y]n
MDL Number: MFCD00081472
CAS Number: 9004-32-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR