Calcium sulfate dihydrate ≥98% NF suitable for use as excipient

Supplier: SPECTRUM CHEMICAL MFG CORP

Synonyms: Plaster of Paris

CA170-2.5KG CA170-500GM
95032-350EA 793.16 CAD
95032-350 95032-352
Calcium sulfate dihydrate ≥98% NF suitable for use as excipient
Calcium sulphate dihydrate

Calcium Sulfate, Dihydrate, NF is used as an excipient for its filler and diluent properties. 


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

Formula: CaSO₄·2H₂O
Density: 2.32 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149625
CAS Number: 10101-41-4
EINECS: 231-900-3
Merck Index: 13,01711

Order Now

Specification Test Results

Assay 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR