Calcium chloride dihydrate ≥99% USP, Ph. Eur., BP, JP

Supplier: Spectrum Chemical Mfg. Corp.
Warning

CA140-50KG CA140-2.5KG CA140-12KG CA140-500GM
89050-408EA 3669.08 CAD
89050-408 95032-246 700001-054 CA95032-248
Calcium chloride dihydrate ≥99% USP, Ph. Eur., BP, JP
Calcium chloride dihydrate

Calcium Chloride, Dihydrate, USP, EP, BP, JP is used in treatment of hypocalcemia, electrolyte depletion, and hyperkalemia. All Spectrum Chemical USP, EP, BP, JP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities

Formula: CaCl₂·2H₂O
Boiling Pt: 1600 °C (1013 hPa)
Melting Pt: 176 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149613
CAS Number: 10035-04-8
EINECS: 233-140-8
Merck Index: 13,01660

Order Now

Specification Test Results

Assay 99% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR