Bromothymol blue ACS

Supplier: Spectrum Chemical Mfg. Corp.

BR154-100GM BR154-25GM BR154-5GM
CA700009-096EA 2951 CAD
CA700009-096 CA700009-100 CA700009-102
Bromothymol blue ACS
Bromothymol blue

Bromothymol Blue, Reagent, ACS is a pH indicator for weak acids and bases. It is mostly used in applications that require measuring substances that would have a relatively neutral pH (near 7), such as managing the pH of pools and fish tanks.


As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Formula: C₂₇H₂₈Br₂O₅S
Melting Pt: 204…202 °C
Storage Temperature: Ambient
MDL Number: MFCD00005872
CAS Number: 76-59-5
EINECS: 200-971-2
Merck Index: 13,01430

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR