1-Butanol NF
Supplier: SPECTRUM CHEMICAL MFG CORP CA
Synonyms:
n-Butanol
, Propyl carbinol
, 1-Butyl alcohol
, Butyl Alcohol
, 1-Hydroxy butane
, n-Butyl Alcohol
B1197-20LT
B1197-1LTCS6
B1197-200LTB
B1197-4LTGL
B1197-500CS6
B1197
B1197-500MLG
B1197-500MLG
B1197-1LT
B1197-200LTB
B1197-4CS4
CA11028-234EA
3611.6
CAD
700002-660
95030-784
89078-644
95030-782
CA11028-234
95030-780
95030-802
700002-658
95030-800
89078-642
95030-786
1-Butanol NF
1-Butanol
Butyl Alcohol, NF is primarily a solvent.
All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).
Formula:
H₃C(CH₂)₂CH₂OH MW: 74.12 g/mol Boiling Pt: 117.6 °C (1013 hPa) Melting Pt: –89 °C Density: 0.811 Flash Pt: 30 °C |
MDL Number:
MFCD00002964 CAS Number: 71-36-3 UN: 1120 ADR: 3,III Merck Index: 13,01539 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...