Nalgene® Color-Coded Low Density Polythylene Wash Bottles, Thermo Scientific
Supplier: Thermo Fisher Scientific
2422-2500 2422-3500 2422-4500 2422-5500
16652-010CS 314.74 CAD
16652-010 16652-012 16652-014 16652-016
Nalgene® Color-Coded Low Density Polythylene Wash Bottles, Thermo Scientific
Bottles Wash Bottles
High-quality squeeze bottles with color-coded closures for easy identification.
  • Color-coded top allows coordination of reagents with industry storage codes
  • Tip can be cut back to provide increased flow
  • Tight fit of draw tube and closure prevents separation when squeezing bottle
  • Flexible LDPE bottle provides soft, ergonomic squeeze
  • Sputter-free, precision stream
  • Bottle and carton are readily recyclable in most communities
  • Manufactured from non-toxic materials
Nalgene® Color-Coded LDPE wash bottles are made from lab quality low density polyethylene (More Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable