Non-Detergent Sulfobetaine (ND SB) 201, G-Biosciences
Supplier: G-Biosciences
DG080 DG081
CA82023-044EA 691.17 CAD
CA82023-044 CA82023-046
Non-Detergent Sulfobetaine (ND SB) 201, G-Biosciences
Protein Purification Protein Purification Resins
NDSB 201 is a zwitterionic compound
Unlike zwitterionic detergents, the hydrophobic group in NDSB 201 is too short to form micelles, even at 1M concentrations. NDSB 201 has been used for purification of proteins and solubilization of protein samples for 2D gel electrophoresis.
Ordering information: Accessories not included.