Non-Ionic Detergent Solutions
Supplier: G-Biosciences
DG015 DG016 DG017 DG018 DG019 DG020 DG021 DG022 DG023 DG024 DG025 DG026 786-519 786-520 786-521 786-522 786-511 786-512 786-517 786-518 786-351 786-352
CA82022-992EA 632.3 CAD
CA82022-992 CA82022-994 CA82022-996 CA82022-998 CA82023-000 CA82023-002 CA82023-004 CA82023-006 CA82023-008 CA82023-010 CA82023-012 CA82023-014 CA95043-486 CA95043-488 CA95043-490 CA95043-492 CA95043-470 CA95043-472 CA95043-482 CA95043-484 CA95057-560 CA95057-562
Non-Ionic Detergent Solutions
Cleaning Agents Detergents and Surface Cleansers
Non-ionic detergents have a hydrophilic head group that is uncharged and are preferred for their ability to break lipid-lipid and lipid-protein interactions.
The non-ionic deteregents are supplied as a general Research Grade, Proteomic Grade (PG) Solutions and 2D-Detergents™. The Proteomic Grade (PG) Solutions have ultra low aldehyde (<100 μM) and peroxide (<50 μM) concentrations to reduce the effects of peroxidase and carbonyl compounds that negatively interact with membrane proteins. The 2D-Detergents™ have low conductivity (<10 μS) and ultra low aldehyde (<100 μM) and peroxide (<50 μM) concentrations.
G-Biosciences Non-Ionic Detergent Solutions include: Triton® X-100, Triton® X-114, Brij® 35, Brij® 58, Tween® 20, Tween® 80, Nonidet® P-40 Substitute, Octyl ß Glucoside, MEGA 8, MEGA 9, MEGA 10, BigCHAP, and Deoxy Big CHAP.
They have limited ability to break protein-protein interactions and are often referred to as non-denaturing detergents and are used to isolate biologically active membrane proteins.
Ordering information: Accessories not included.