TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, 2D-Detergent™, purified

Supplier: G-Biosciences

Synonyms: Polyethyleneglycol-[4-(1,1,3,3-tetramethylbutyl)phenyl]-ether, (p-tert-Octylphenoxy)polyethoxyethanol, Octoxynol, Polyoxyethylene(10) Octylphenyl Ether, t-Octylphenoxypolyethoxyethanol, O-[4-(1,1,3,3-Tetramethylbutyl)phenoxy]polyethoxyethanol

DG907 DG908
CA95057-622EA 603.92 CAD
CA95057-622 CA95057-624
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether) solution 10%, 2D-Detergent™, purified
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)

Many commercial grade detergents contain elevated levels of sulfhydryl oxidizing agents, peroxides, salts and carbonyl compounds. 2-D Detergent™ Octylphenolpoly(ethyleneglycolether)x (Triton® X-100 substitute), 10% aqueous solution.



2D-Detergent™ solutions contain reduced peroxides and carbonyl compounds. In addition, the detergents have less than 15µS conductivity. These detergents are offered as 10% aqueous solutions, sealed under inert gas and are suitable for all protein applications, including 2D-electrophoresis. These non-ionic detergents are suitable for isolating membrane-protein complexes. The aldehyde and peroxide levels are <50µM and <10µM respectively with a conductivity of <15µS.


The proteins that are isolated with these detergents are highly susceptible to contaminating peroxides and carbonyls. The peroxides will oxidize proteins and the carbonyl groups will form Schiff's bases with the proteins that interfere with a protein's structure.


Ordering information: Accessories not included.

Melting Pt: 6
Flash Pt: 251
MDL Number: MFCD00132505
CAS Number: 9002-93-1
Merck Index: 13,06793

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR