Proteomic Grade Detergent Solutions
Supplier: G-Biosciences
DG001 DG002 DG005 DG006 DG007 DG008 DG009 DG010 DG012 DG013 DG521
CA82022-966EA 656.44 CAD
CA82022-966 CA82022-968 CA82022-974 CA82022-976 CA82022-978 CA82022-980 CA82022-982 CA82022-984 CA82022-986 CA82022-988 CA95057-526
Proteomic Grade Detergent Solutions
Cleaning Agents Detergents and Surface Cleansers
Many commercial grade detergents contain elevated levels of sulfhydryl oxidizing agents, peroxides, salts and carbonyl compounds.
Our Proteomic Grade Detergent Solutions contain reduced peroxides and carbonyl compounds. In addition, the detergents have less than 50 μS conductivity. These detergents are offered as 10% aqueous solutions, sealed under inert gas and are suitable for protein applications. These non-ionic detergents are suitable for isolating membrane-protein complexes.
G-Biosciences Proteomic Grade Detergent Solutions include: Triton® X-100, Triton® X-114, Brij® 35, Brij® 58
More Product Information