Tube-O-Reactor™ for Protein Cross Linking, Coupling and Modification, G-Biosciences
Supplier: G-Biosciences
786-027-4K 786-027-8K 786-027-15K 786-024-15K 786-024-4K 786-024-8K
CA82021-244EA 547.15 CAD
CA82021-244 CA82021-246 CA82021-248 CA95043-370 CA95043-372 CA95043-374
Tube-O-Reactor™ for Protein Cross Linking, Coupling and Modification, G-Biosciences
Tubes Dialysis Tubes
Tube-O-Reactor™ is a system that allows all the key steps of cross linking, coupling and modification of proteins and/or nucleic acids to be performed in a single tube.
Most of the above reactions involve three main steps, including: the equilibration of reaction conditions for optimized reactions, the subsequent reaction with target agents (i.e. cross linkers and labels), and the removal of unreacted agents and by-products.
The Tube-O-Reactor™ system is available in three MWCO sizes, 4 kDa, 8 kDa and 15 kDa. Tube-O-Reactor™ is supplied as a Micro kit for sample sizes of 20 to 250 μl and a Medi size for samples of 0.2 to 2 ml. Each Tube-O-Reactor™ is suitable for five reactions, depending on sample volumes, and is supplied with: Five Medi or five Micro Tube-O-DIALYZER™, five Floats for each size of Tube-O- DIALYZER™, five Micro Dialysis Reaction Chambers,and glass balls.
This minimizes the risk of sample loss, of experimental time and of hands-on phases.
Ordering information: Accessories not included.