BIOPURE 15 Water Purification System with Booster Pump
Supplier: ELGA LabWater
BIOPURE 15
MB015BPM1
CA89204-162EA 12162.42 CAD
CA89204-162
BIOPURE 15 Water Purification System with Booster Pump
Water Purification Systems Pure and Ultra Pure Water Systems
The Biopure 15 is ideal for ENT and dental decontamination.
  • Dispense Rate up to 2.0 LPM
  • Make Up Flow Rate 5 LPH (360 L/day)
  • Water Quality ype II
High Performance Water Purification System designed to feed washer disinfectors and endoscope reprocessors. Feed water booster pump included.
It can directly supply 2 L/min of compliant purified water directly to a wide range of applications including washer disinfectors, autoclaves, Ultra sonic baths, ENT Automated Endoscope Reprocessors, etc. The Biopure 15 can produce up to 15 L/hr of purified water.