CENTRA® Water Purification Systems, ELGA LabWater
Supplier: ELGA CA
CN120RDM1-115-US CN200RDM1-115-US CNHFRR2M1-230-US CNXXXRDM1-115-US
CA89204-144EA 38410.04 CAD
CA89204-144 CA89204-146 CA89204-148 CA89204-150
CENTRA® Water Purification Systems, ELGA LabWater
Water Purification Systems Pure and Ultra Pure Water Systems
The CENTRA has revolutionised the way that large volumes of pure water are produced, stored and distributed
  • Compact design
  • Reliable, continual supply of pure water
  • Unique access controls, leak detection systems and full AV alarms
  • Optional building management system (BMS) connectivity
  • Optimized inorganic water quality
  • In-line purification technologies
The compact design provides more flexible installation options for new buildings and refurbishments. The CENTRA’s small footprint means that it can be placed closer to the laboratory, avoiding the negative cost and design implications of long pipe work loops. Recirculated water is UV treated, filtered and (if fitted) can be improved through deionization.
Instead of a central lab system, comprising of an untidy grouping of components and pipework, you can now have just one integrated system box.
Accessories information: Additional accessories are available; contact your Avantor representative for more information.
Ordering information: For CENTRA R200 models, the booster pump module for the CENTRA® High Flow needs to be ordered separately. A pretreatment pack and service DI tank may be required to produce Type II quality water. For a distribution loop up to 1,000 feet, the booster pump module (89204-256) will have to be ordered separately. The CENTRA-RDS is a storage and recirculating distribution system designed to work with other CENTRA or PURELAB systems or existing purification installations. For CENTRA R120, the starter kit and DI pack need to be ordered separately. Optional purchases include a .2 micron filter and e-cartridge. The system can operate as an RO or RO/DI; depending on the applications for which the system is used, filters might need to be purchased.
Please speak to your VWR representative to determine the correct unit for your requirements.