PURELAB® flex 1 and 2 Water Purification Systems, ELGA LabWater
Supplier: ELGA CA
PF1XXXXM1-US PF2XXXXM1-US SK-FLEX1-KIT SK-FLEX2-KIT SK-PF1XXXXM1-KIT SK-PF2XXXXM1-KIT SK-CSPFLEX-A SK-CSPFLEX-B SK-LPMAFLEX-12 SK-LPMAFLEX-12
CA89204-088EA 8761.85 CAD
CA89204-088 CA89204-092 89221-838 89221-840 CA89221-838 CA89221-840 76516-398 76516-400 76670-678 CA76670-678
PURELAB® flex 1 and 2 Water Purification Systems, ELGA LabWater
Water Purification Systems Pure and Ultra Pure Water Systems
The PURELAB® flex range is designed to deliver accuracy, flexibility and ease-of-use. The award-winning system provides perfect water purity for analytical and life science applications which require RO type III water up to ultrapure type I (18.2 MΩ.cm) water. It allows focus on routine test work without concern about the water quality affecting test results.
  • Customized settings
  • Easy access to the consumables through the front door panels
  • Maintenance time is reduced, with less disruption to your work
  • Download all of the data to USB for system performance
More Product Information
Order Now
SPECIFICATIONS
Model PURELAB® flex 1 PURELAB® flex 2 PURELAB® flex 1 PURELAB® flex 2 PURELAB® flex 1 PURELAB® flex 2
Average daily needs <10 L <10 L <10 L <10 L <10 L <10 L
Environmentally Preferable
Flow rate max. 2.0 L/min max. 2.0 L/min max. 2.0 L/min max. 2.0 L/min max. 2.0 L/min max. 2.0 L/min
TOC content Dependent on feedwater <5 ppb Dependent on feedwater <5 ppb Dependent on feedwater <5 ppb
Inorganics @ 25°C 18.2 MΩ.cm with DI pack 18.2 MΩ.cm 18.2 MΩ.cm with DI pack 18.2 MΩ.cm 18.2 MΩ.cm with DI pack 18.2 MΩ.cm
Bacteria <0.1 CFU/ml with POU filter fitted <0.1 CFU/ml with POU filter fitted <0.1 CFU/ml with POU filter fitted <0.1 CFU/ml with POU filter fitted <0.1 CFU/ml with POU filter fitted <0.1 CFU/ml with POU filter fitted
Endotoxin <0.001 EU/mL with POU Biofilter fitted <0.001 EU/mL with POU Biofilter fitted <0.001 EU/ml with POU Biofilter fitted <0.001 EU/ml with POU Biofilter fitted <0.001 EU/mL with POU Biofilter fitted <0.001 EU/mL with POU Biofilter fitted
Electrical 100-240V AC, 50-60Hz 100-240V AC, 50-60Hz 100-240V AC, 50-60Hz 100-240V AC, 50-60Hz 100-240V AC, 50-60Hz 100-240V AC, 50-60Hz