Nalgene® Invitro™ Biotainer Bottles, Sterile, HDPE, Thermo Scientific
Supplier: Thermo Fisher Scientific
3751-42 3750-24 3751-24
82028-492CS 975.81 CAD
82028-492 82028-494 82028-496
Nalgene® Invitro™ Biotainer Bottles, Sterile, HDPE, Thermo Scientific
Bottles Plastic Bottles
Nalgene® InVitro Biotainer bottles reduce the risk of carryover contamination and eliminate the cost of cleaning, sterilizing and its associated validations.
  • The bottles are supplied sterile and ready to use, and offer excellent chemical resistance
They are made from HDPE with a silicone-lined polypropylene 38 mm closure. They feature printed graduations to 3000 ml, with a 3900 ml brim capacity and a space-saving square shape with a convenient handle.
Certifications: Meet USP 87, 88. Lot tested for pyrogenicity.
Ordering information: 3750-24More Product Information