Nalgene® PETG Certified Clean Containers, Thermo Scientific
Supplier: Thermo Fisher Scientific
382019-0030 382019-0060 382019-0125 382019-0250 382019-0500 382019-1000 382019-2000
89093-194CS 527.09 CAD
89093-194 89093-196 89093-198 89093-200 89093-202 89093-204 89093-206
Nalgene® PETG Certified Clean Containers, Thermo Scientific
Bottles Plastic Bottles
Nalgene® PETG Certified Clean Containers are suitable for pharmaceutical and biotechnology applications requiring the processing and storage of bulk intermediates and critical reagents such as vaccine and protein therapeutic preparations.
The packaging is designed for cleanroom applications. The bottle and closure are secured with heat-shrink bands providing a tamper-evident seal and then packaged in cleanroom grade heat-sealed bags to assure product integrity. They are then gamma irradiated to provide a 10⁻⁶ sterility assurance level (SAL). Bottles meet the requirements for USP Class VI,More Product Information