Nalgene® Disposable Erlenmeyer Flasks, PETG, Sterile, Baffle Bottom, Thermo Scientific
Supplier: Thermo Fisher Scientific
4113-0250 4113-0500 4113-2000 4113-0125 4113-1000 4113-2800
73520-402CS 295.86 CAD
73520-402 73520-404 73520-406 73520-466 73520-468 82031-374
Nalgene® Disposable Erlenmeyer Flasks, PETG, Sterile, Baffle Bottom, Thermo Scientific
Flasks Erlenmeyer Flasks
Graduated flasks are made of light, crystal-clear PETG plastic with easy-to-open polyethylene screw closures.
  • Disposing of flasks reduces chance of cross-contamination
  • Non-pyrogenic, non-cytotoxic
  • Sterilized to SAL 10-6
The leak-proof high-density polyethylene screw closures open to vent with 1/4 turn.
Ideal for shaker and suspension cell culture, media preparation, or storage
Packaging: Individually packaged for easy storage.
Order Now
SPECIFICATIONS
Environmentally Preferable