VWR® Protective Eyewear with Wraparound Lens
Supplier: VWR International
89187-986 76329-622 76329-624 76329-626
89187-986CS 779.44 CAD
89187-986 76329-622 76329-624 76329-626
VWR® Protective Eyewear with Wraparound Lens
Safety Glasses
VWR® protective eyewear with wraparound lens offers stylish and secure personal protection, with an array of comfort features to assist the wearer throughout extended use.
  • Fit comfortably with half-mask respirators
  • Impact-resistant, scratch-resistant polycarbonate lens
  • Provides 99.9% UV protection
Super lightweight wraparound eyewear for men and women combines a sleek appearance with rugged durability. Eyewear features rubberized temple tips and an integrated nose piece for extra comfort.
Certifications: ANSI Z87.1 certified.
Order Now
SPECIFICATIONS
Environmentally Preferable