VWR® Visitor Specs
Supplier: VWR
VWR®
5120
89187-988CS 645.49 CAD
89187-988
VWR® Visitor Specs
Safety Glasses
VWR® visitor specs offer stylish and secure protection with an array of comfort features to enable extended use.
  • Designed to fit comfortably over most prescription glasses
  • Wraparound styling offers unobstructed peripheral vision
  • Contoured protective brow guard and molded-in comfort nose bridge
  • Impact-resistant, scratch-resistant polycarbonate lenses
  • Provides 99.9% UV protection
Molded from 100% polycarbonate material, this impact-resistant eyewear features wide vision wrap-around styling for unobstructed peripheral vision, a contoured protective brow-guard which helps prevent particles from entering the brow area, and a molded-in comfort nose bridge that provides a comfortable fit on the nose. Specs have a scratch-resistant coating on the lens, and fit over most prescription glasses.
Certifications: ANSI Z87.1 certification.