Allegra® X-30R Series Benchtop Centrifuges, Beckman Coulter
Supplier: Beckman Coulter
B05796 B05802 B06314 B06320 B08536 B08538 B08540 B08542 B08545 B08547 B08705 B08708 B05796 B05802 B06314 B06320 B08536 B08538 B08540 B08542 B08545 B08547 B08705 B08708
BKB05796EA 18836.3 CAD
BKB05796 BKB05802 BKB06314 BKB06320 BKB08536 BKB08538 BKB08540 BKB08542 BKB08545 BKB08547 BKB08705 BKB08708 CABKB05796 CABKB05802 CABKB06314 CABKB06320 CABKB08536 CABKB08538 CABKB08540 CABKB08542 CABKB08545 CABKB08547 CABKB08705 CABKB08708
Allegra® X-30R Series Benchtop Centrifuges, Beckman Coulter
Centrifuges Benchtop Centrifuges
The Allegra® X-30R series benchtop centrifuges offer high performance and versatility in a compact design.
  • Cell Culture Package includes centrifuge and rotor SX4400
  • Microplate Application Package includes centrifuge and microplate rotor S606
  • Molecular Performance Application Package includes centrifuge and fixed-angle rotor F2402H
  • Clinical Applications Package includes centrifuge, rotor SX4400, and adapters
The Allegra® X-30R refrigerated models offer a g-force max of 29,756xg (18,000rpm) for fixed angle rotors and 4,255xg (4700rpm) for swing bucketMore Product Information