Corning® Disposable Tubing Set for Use with Glass Bottle, Corning
Supplier: Corning
10042 10043
89184-630CS 443.47 CAD
89184-630 89184-632
Corning® Disposable Tubing Set for Use with Glass Bottle, Corning
Tubing Transfer Tubing
Chemically resistant, heat sealable flexible tubing. Accommodates both 2 L and 4 L bottles. Male MPC connector with cap.
  • PureFit® tube clamp system
  • Autoclavable