Anti-p21 WAF1 (EA10) Mouse Monoclonal Antibody

Supplier: MilliporeSigma
OP64-100UG OP64-20UG
CA80001-388EA 1150.53 CAD
CA80001-388 CA82602-450
Anti-p21 WAF1 (EA10) Mouse Monoclonal Antibody
Antibodies
Primary mouse Anti-p21 WAF1 (Ab-1) (EA10) reacts with human.

Type: Primary
Antigen: p21 WAF1 (EA10)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR