Anti-TBK1 Mouse Monoclonal Antibody

Supplier: EMD Millipore
AM76-100UG
CA80600-622EA 1216.22 CAD
CA80600-622
Anti-TBK1 Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-TBK1 (108A429) Reacts with Human

Type: Primary
Antigen: TBK1
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR