Anti-SV40 T Mouse Monoclonal Antibody

Supplier: EMD Millipore
DP01-100UG
CA80000-348EA 1090.97 CAD
CA80000-348
Anti-SV40 T Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-SV40 T Antigen (Ab-1) (PAb419) Reacts with SV40 INF

Type: Primary
Antigen: SV40 T Antigen (Ab-1) (PAb419)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: SV40 INF
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR