Anti-GSK3B Mouse Monoclonal Antibody

Supplier: EMD Millipore
361527-1SET
CA80601-192EA 1561.21 CAD
CA80601-192
Anti-GSK3B Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-GSK3 beta (pSer 9 ) (2D3) Reacts with Human, Mouse

Type: Primary
Antigen: GSK3 beta (pSer 9 ) (2D3)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR