Anti-PDGFR beta Rabbit Polyclonal Antibody

Supplier: EMD Millipore
PK1008-50UL
CA82602-476EA 905.53 CAD
CA82602-476
Anti-PDGFR beta Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-PDGFR beta(pTyr 751 ) Reacts with Human, Mouse, Rat

Type: Primary
Antigen: PDGFR beta(pTyr 751 )
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR