Anti-Neurofilament H (SMI-31) Mouse Monoclonal Antibody

Supplier: EMD Millipore
NE1022-100UL
CA82602-440EA 1342.13 CAD
CA82602-440
Anti-Neurofilament H (SMI-31) Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-Neurofilament H (SMI-31) Reacts with Chicken, Mammalia, Xenopus Laveus

Type: Primary
Antigen: Neurofilament H (SMI-31)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Chicken, Mammals, Xenopus Laveus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR