Anti-NF-kappa B (p65, RelA) Rabbit Polyclonal Antibody

Supplier: EMD Millipore
PC138-100UL
CA80001-544EA 1342.13 CAD
CA80001-544
Anti-NF-kappa B (p65, RelA) Rabbit Polyclonal Antibody
Antibodies
Primary Rabbit Anti-NF-kappa B (p65, RelA) (Ab-2) Reacts with Human

Type: Primary
Antigen: NF-kappa B (p65, RelA) (Ab-2)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR