Anti-p53 (Ab-6) Mouse Monoclonal Antibody [clone: DO-1]

Supplier: MilliporeSigma
OP43-100UG OP43L-100UG OP43-20UG
CA80001-350EA 1214.07 CAD
CA80001-350 CA80001-356 CA80016-902
Anti-p53 (Ab-6) Mouse Monoclonal Antibody [clone: DO-1]
Antibodies
This Anti-p53 (Ab-6) (Pantropic) Mouse mAb (DO-1) is validated for use in frozen sections, immunoblotting, ICC, IP, paraffin sections for the detection of p53 (Ab-6) (Pantropic).

Type: Primary
Antigen: p53 (Ab-6)
Clonality: Monoclonal
Clone: DO-1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse, Rat, Cat, Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR