Anti-CREBBP Mouse Monoclonal Antibody

Supplier: EMD Millipore
NA46-100UG
CA80000-886EA 1091.38 CAD
CA80000-886
Anti-CREBBP Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-p300 (NM-11) Reacts with Human, Rat

Type: Primary
Antigen: p300 (NM-11)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR