Anti-ITGA3 Mouse Monoclonal Antibody

Supplier: EMD Millipore
CP11L-100UG
CA80000-216EA 1133.1 CAD
CA80000-216
Anti-ITGA3 Mouse Monoclonal Antibody
Antibodies
Primary Mouse Anti-Integrin alpha 3 (P1B5) Reacts with Human

Type: Primary
Antigen: Integrin alpha 3 (P1B5)
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR